Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244531.1 | NP_001231460.1 | 6 - 7 | 748 | 82 | |
GenBank mRNA | AK235228.1 | - | 6 - 7 | 790 | 82 | |
AK235800.1 | - | 6 - 7 | 789 | 82 | ||
AK348125.1 | - | 6 - 7 | 750 | 82 | ||
AK351883.1 | - | 6 - 7 | 748 | 82 | ||
AK392207.1 | - | 6 - 7 | 778 | 82 | ||
AK392777.1 | - | 6 - 7 | 758 | 82 | ||
AK398410.1 | - | 5 - 6 | 490 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Sus scrofa Inventoried Probe spans exons |