Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFT2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244534.1 | NP_001231463.1 | 2 - 3 | 224 | 74 | |
GenBank mRNA | AK234386.1 | - | 2 - 3 | 212 | 74 | |
AK392532.1 | - | 2 - 3 | 201 | 74 | ||
AK400085.1 | - | 2 - 3 | 227 | 74 | ||
BW981531.1 | - | 2 - 3 | 224 | 74 | ||
EW622209.2 | - | 2 - 3 | 237 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Sus scrofa Probe spans exons |