Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFT2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244534.1 | NP_001231463.1 | 3 - 4 | 303 | 111 | |
GenBank mRNA | AK234386.1 | - | 3 - 4 | 291 | 111 | |
AK392532.1 | - | 3 - 4 | 280 | 111 | ||
AK400085.1 | - | 3 - 4 | 306 | 111 | ||
EW622209.2 | - | 3 - 4 | 316 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |