Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KAZALD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244551.1 | NP_001231480.1 | 1 - 2 | 119 | 103 | |
GenBank mRNA | AK350351.1 | - | 1 - 2 | 119 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Sus scrofa Probe spans exons |