Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRFBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244566.1 | NP_001231495.1 | 3 - 4 | 240 | 73 | |
XM_021079835.1 | XP_020935494.1 | 342 | 73 | |||
GenBank mRNA | AK234890.1 | - | 3 - 4 | 240 | 73 | |
AK390530.1 | - | 3 - 4 | 336 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Sus scrofa Probe spans exons |