Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244582.1 | NP_001231511.1 | 1 - 2 | 268 | 98 | |
GenBank mRNA | AK395833.1 | - | 1 - 2 | 268 | 98 | |
FS692142.1 | - | 1 - 2 | 269 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |