Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244622.1 | NP_001231551.1 | 7 - 8 | 1283 | 110 | |
GenBank mRNA | AK349825.1 | - | 7 - 8 | 1283 | 110 | |
AK397166.1 | - | 7 - 8 | 1286 | 110 | ||
AK397400.1 | - | 7 - 8 | 1280 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |