Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF3I | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244624.1 | NP_001231553.1 | 2 - 3 | 150 | 105 | |
GenBank mRNA | AK232367.1 | - | 2 - 3 | 150 | 105 | |
AK394444.1 | - | 2 - 3 | 153 | 105 | ||
AK397989.1 | - | 2 - 3 | 140 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |