Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C5H12orf45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244659.1 | NP_001231588.1 | 1 - 2 | 175 | 102 | |
XM_021091093.1 | XP_020946752.1 | 170 | 102 | |||
GenBank mRNA | AK237454.1 | - | 1 - 2 | 122 | 102 | |
AK344341.1 | - | 1 - 2 | 175 | 102 | ||
AY609857.1 | - | 1 - 2 | 115 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Sus scrofa Probe spans exons |