Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC28A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244669.1 | NP_001231598.1 | 5 - 6 | 688 | 90 | |
GenBank mRNA | AK231417.1 | - | 5 - 6 | 688 | 90 | |
AK395072.1 | - | 5 - 6 | 702 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Sus scrofa Probe spans exons |