Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244786.1 | NP_001231715.1 | 2 - 3 | 386 | 63 | |
GenBank mRNA | AK231845.1 | - | 2 - 3 | 388 | 63 | |
AK239323.1 | - | 2 - 3 | 393 | 63 | ||
AY610052.1 | - | 2 - 3 | 375 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |