Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244786.1 | NP_001231715.1 | 3 - 4 | 500 | 100 | |
GenBank mRNA | AK231845.1 | - | 3 - 4 | 502 | 100 | |
AY610052.1 | - | 3 - 4 | 489 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Sus scrofa Probe spans exons |