Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FADS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244792.1 | NP_001231721.1 | 5 - 6 | 912 | 65 | |
XM_021066101.1 | XP_020921760.1 | 883 | 65 | |||
GenBank mRNA | AK238564.1 | - | 5 - 6 | 912 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Sus scrofa Probe spans exons |