Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244823.1 | NP_001231752.1 | 2 - 3 | 275 | 69 | |
XM_005669173.3 | XP_005669230.1 | 423 | 69 | |||
GenBank mRNA | AK344929.1 | - | 2 - 3 | 275 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Sus scrofa Probe spans exons |