Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCHF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244831.1 | NP_001231760.1 | 2 - 3 | 465 | 69 | |
GenBank mRNA | AK235372.1 | - | 2 - 3 | 465 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Sus scrofa Probe spans exons |