Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001244833.1 | NP_001231762.1 | 4 - 5 | 458 | 95 | |
GenBank mRNA | AK235370.1 | - | 4 - 5 | 458 | 95 | |
AK390254.1 | - | 4 - 5 | 481 | 95 | ||
AK392244.1 | - | 3 - 4 | 207 | 95 | ||
AK399104.1 | - | 3 - 4 | 225 | 95 | ||
AY609557.1 | - | 4 - 5 | 429 | 95 | ||
KP735785.1 | - | 4 - 5 | 487 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Sus scrofa Probe spans exons |