Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DBNDD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243672.1 | NP_001230601.1 | 2 | 319 | 87 | |
GenBank mRNA | AK239121.1 | - | 2 | 319 | 87 | |
AK239444.1 | - | 2 | 309 | 87 |
Gene Symbol: SYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005672967.3 | XP_005673024.1 | 355 | 87 | ||
GenBank mRNA | AK393775.1 | - | 2 | 327 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Sus scrofa Both primers and probe map within a single exon |