Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003134142.5 | XP_003134190.2 | 678 | 59 | ||
XM_013984919.2 | XP_013840373.2 | 490 | 59 | |||
GenBank mRNA | AK234885.1 | - | 3 - 4 | 590 | 59 | |
AK390720.1 | - | 3 - 4 | 495 | 59 | ||
AK391379.1 | - | 3 - 4 | 505 | 59 | ||
AK391464.1 | - | 3 - 4 | 544 | 59 | ||
AK391491.1 | - | 3 - 4 | 532 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Sus scrofa Probe spans exons |