Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF359521.1 | - | 2 - 3 | 396 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Vitis vinifera Probe spans exons |