Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MADS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF373601.1 | - | 3 - 4 | 366 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Vitis vinifera Probe spans exons |