Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ237988.1 | - | 6 - 7 | 459 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Vitis vinifera Probe spans exons |