Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIP55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ237992.1 | - | 4 - 5 | 1371 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Vitis vinifera Probe spans exons |