Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ490336.1 | - | 10 - 11 | 1761 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Vitis vinifera Probe spans exons |