Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY156048.1 | - | 1 - 2 | 396 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |