Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VFUL-L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY275713.1 | - | 4 - 5 | 388 | 99 | |
AY538747.1 | - | 4 - 5 | 417 | 99 | ||
GU133635.1 | - | 3 - 4 | 334 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Vitis vinifera Probe spans exons |