Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY461710.1 | - | 10 - 11 | 993 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Vitis vinifera Probe spans exons |