Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY861386.1 | - | 2 - 3 | 246 | 97 | |
DQ017393.1 | - | 2 - 3 | 220 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |