Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MADS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ059750.1 | - | 4 - 5 | 453 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Vitis vinifera Probe spans exons |