Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY780886.1 | - | 5 - 6 | 838 | 105 | |
X75964.1 | - | 5 - 6 | 879 | 105 | ||
Y11749.1 | - | 5 - 6 | 883 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |