Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | DQ979341.1 | - | 5 - 6 | 461 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Vitis vinifera Probe spans exons |