Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU447172.1 | - | 5 - 6 | 657 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Vitis vinifera Probe spans exons |