Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLO12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | EU591730.1 | - | 2 - 3 | 121 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Vitis vinifera Probe spans exons |