Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ809943.1 | - | 8 - 9 | 1375 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Vitis vinifera Probe spans exons |