Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIPK09 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | FJ901237.1 | - | 8 - 9 | 758 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Vitis vinifera Probe spans exons |