Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrpb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001198.1 | NP_001001198.1 | 2 - 3 | 230 | 97 | |
XM_018097689.1 | XP_017953178.1 | 2 - 3 | 279 | 97 | ||
GenBank mRNA | BC067325.1 | - | 2 - 3 | 230 | 97 | |
BC076700.1 | - | 2 - 3 | 257 | 97 | ||
CT030073.3 | - | 2 - 3 | 228 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |