Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001205.1 | NP_001001205.1 | 7 - 8 | 788 | 101 | |
XM_018097691.1 | XP_017953180.1 | 6 - 7 | 1469 | 101 | ||
GenBank mRNA | BC067310.1 | - | 7 - 8 | 788 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |