Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ap2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001209.2 | NP_001001209.1 | 10 - 11 | 1380 | 94 | |
XM_012960782.2 | XP_012816236.1 | 10 - 11 | 1556 | 94 | ||
XM_012960783.2 | XP_012816237.1 | 10 - 11 | 1556 | 94 | ||
XM_012960784.2 | XP_012816238.1 | 10 - 11 | 1555 | 94 | ||
GenBank mRNA | BC067918.2 | - | 10 - 11 | 1380 | 94 | |
CR760468.2 | - | 3 - 4 | 362 | 94 | ||
CR848593.2 | - | 10 - 11 | 1357 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |