Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001216.1 | NP_001001216.1 | 1 - 2 | 488 | 59 | |
GenBank mRNA | BC067972.1 | - | 1 - 2 | 488 | 59 | |
CR760274.1 | - | 1 - 2 | 373 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |