Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: llph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001225.1 | NP_001001225.1 | 2 - 3 | 293 | 98 | |
GenBank mRNA | BC067940.1 | - | 2 - 3 | 293 | 98 | |
CR760795.2 | - | 2 - 3 | 261 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |