Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: irf2bp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001988.1 | NP_001001988.1 | 1 - 2 | 1484 | 77 | |
GenBank mRNA | BC063909.1 | - | 1 - 2 | 915 | 77 | |
BC075565.1 | - | 1 - 2 | 1484 | 77 | ||
CT010551.2 | - | 1 - 2 | 1107 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |