Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002902.1 | NP_001002902.1 | 9 - 10 | 772 | 75 | |
XM_012956694.2 | XP_012812148.1 | 10 - 11 | 75 | |||
XM_018091156.1 | XP_017946645.1 | 9 - 10 | 75 | |||
GenBank mRNA | BC074572.1 | - | 9 - 10 | 772 | 75 | |
CR762304.2 | - | 8 - 9 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |