Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fut10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002903.1 | NP_001002903.1 | 1 - 2 | 71 | 106 | |
XM_012958786.2 | XP_012814240.1 | 3 - 4 | 788 | 106 | ||
GenBank mRNA | AJ784815.1 | - | 1 - 2 | 71 | 106 | |
BC123051.1 | - | 3 - 4 | 341 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |