Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004792.1 | NP_001004792.1 | 6 - 7 | 750 | 74 | |
XM_012970284.2 | XP_012825738.1 | 6 - 7 | 1148 | 74 | ||
XM_018097075.1 | XP_017952564.1 | 6 - 7 | 810 | 74 | ||
GenBank mRNA | BC074528.1 | - | 6 - 7 | 750 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |