Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004792.1 | NP_001004792.1 | 7 - 8 | 879 | 70 | |
XM_012970284.2 | XP_012825738.1 | 7 - 8 | 1277 | 70 | ||
XM_018097075.1 | XP_017952564.1 | 7 - 8 | 939 | 70 | ||
GenBank mRNA | BC074528.1 | - | 7 - 8 | 879 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |