Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rprd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004796.1 | NP_001004796.1 | 2 - 3 | 340 | 58 | |
XM_012971622.2 | XP_012827076.1 | 2 - 3 | 378 | 58 | ||
GenBank mRNA | BC074536.1 | - | 2 - 3 | 340 | 58 | |
CT025363.2 | - | 2 - 3 | 297 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |