Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gorasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004805.1 | NP_001004805.1 | 5 - 6 | 661 | 85 | |
GenBank mRNA | BC074563.1 | - | 5 - 6 | 661 | 85 | |
CR855824.1 | - | 5 - 6 | 647 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |