Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif1ax | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004814.1 | NP_001004814.1 | 6 - 7 | 671 | 86 | |
GenBank mRNA | BC074588.1 | - | 6 - 7 | 671 | 86 | |
CR760442.2 | - | 6 - 7 | 471 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |