Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp1r11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004815.1 | NP_001004815.1 | 2 - 3 | 260 | 87 | |
GenBank mRNA | BC074590.1 | - | 2 - 3 | 260 | 87 | |
CR760958.2 | - | 2 - 3 | 241 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |