Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ran | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004829.1 | NP_001004829.1 | 4 - 5 | 489 | 101 | |
XM_012959006.2 | XP_012814460.1 | 4 - 5 | 291 | 101 | ||
XM_012959012.2 | XP_012814466.1 | 4 - 5 | 439 | 101 | ||
XM_018091941.1 | XP_017947430.1 | 4 - 5 | 390 | 101 | ||
GenBank mRNA | BC074619.1 | - | 4 - 5 | 489 | 101 | |
CR761555.2 | - | 3 - 4 | 254 | 101 | ||
CR761725.2 | - | 3 - 4 | 262 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |