Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dscc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004834.1 | NP_001004834.1 | 6 - 7 | 777 | 66 | |
GenBank mRNA | BC074630.1 | - | 6 - 7 | 777 | 66 | |
CR760288.2 | - | 6 - 7 | 784 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |